missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CKIP-1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 168.00
Specifications
| Antigen | CKIP-1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
CKIP-1 Polyclonal specifically detects CKIP-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| CKIP-1 | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| NP_057358 | |
| 51177 | |
| Antibody was raised against an 18 amino acid synthetic peptide from near the carboxy terminus of human CKIP1. The immunogen is located within the last 50 amino acids of CKIP1. Amino Acid Squence: TPDSHLRQTTPHSQYKKS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 2810052M02Rik, Casein kinase 2-interacting protein 1, C-Jun-binding protein, CK2 interacting protein 1; HQ0024c protein, CK2-interacting protein 1, CKIP1, CKIP-1PH domain-containing family O member 1, JBP, OC120, Osteoclast maturation-associated gene 120 protein, pleckstrin homology domain containing, family O member 1, pleckstrin homology domain-containing family O member 1 | |
| PLEKHO1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title