missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CKS1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92480-0.02ml
This item is not returnable.
View return policy
Description
CKS1 Polyclonal antibody specifically detects CKS1 in Human samples. It is validated for Western Blot
Specifications
| CKS1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CDC28 protein kinase 1, CDC28 protein kinase 1B, CDC28 protein kinase regulatory subunit 1B, cell division control protein CKS1, CKS-1, CKS1CDC2-associated protein CKS1, ckshs1, cyclin-dependent kinases regulatory subunit 1, NB4 apoptosis/differentiation related protein, PNAS-143, PNAS-16, PNAS-18 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-79 of human CKS1B (NP_001817.1). MSHKQIYYSDKYDDEEFEYRHVMLPKDIAKLVPKTHLMSESEWRNLGVQQSQGWVHYMIHEPEPHILLFRRPLPKKPKK | |
| 0.02 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers | |
| 1163 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction