Learn More
Invitrogen™ CKS2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5104525
Product Code. 16660975
  • € 493.00 / 100µL
Description
Antibody detects endogenous levels of total CKS2.
CKS2 protein binds to the catalytic subunit of the cyclin dependent kinases and is essential for their biological function. The CKS2 mRNA is found to be expressed in different patterns through the cell cycle in HeLa cells, which reflects specialized role for the encoded protein. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CKS2
Polyclonal
Unconjugated
Cks2
1110038L14Rik; CDC28 protein kinase 2; CDC28 protein kinase regulatory subunit 2; CKS1(S. cerevisiae Cdc28/Cdc2 kinase subunit) homolog-2; CKS2; CKS-2; CKSHS2; Cyclin-dependent kinases regulatory subunit 2; RGD1562047; RP11-8C21.2
Rabbit
Affinity Chromatography
RUO
1164, 66197
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P33552, P56390
Cks2
A synthesized peptide derived from human CKS2(Accession P33552), corresponding to amino acid residues R20-R70.