missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CL-K1/COLEC11 Antibody (148), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90474CL1
This item is not returnable.
View return policy
Description
CL-K1/COLEC11 Monoclonal antibody specifically detects CL-K1/COLEC11 in Mouse samples. It is validated for ELISA
Specifications
| CL-K1/COLEC11 | |
| Monoclonal | |
| CoraFluor 1 | |
| CLK1, CL-K1, CL-K1-I, CL-K1-II, CL-K1-IIa, CL-K1-IIb, collectin kidney I, Collectin kidney protein 1, collectin sub-family member 11, collectin-11, DKFZp686N1868, MGC3279 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CL-K1/COLEC11 (Uniprot#: Q3SXB8-1; Met1-Leu272). | |
| 0.1 mL | |
| Cell Biology | |
| 78989 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 148 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction