missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CL-K1/COLEC11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | CL-K1/COLEC11 |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18697222
|
Novus Biologicals
NBP2-98233-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18638654
|
Novus Biologicals
NBP2-98233-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CL-K1/COLEC11 Polyclonal antibody specifically detects CL-K1/COLEC11 in Mouse samples. It is validated for ELISASpecifications
| CL-K1/COLEC11 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| 0.2 um filtered solution in PBS | |
| 78989 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CLK1, CL-K1, CL-K1-I, CL-K1-II, CL-K1-IIa, CL-K1-IIb, collectin kidney I, Collectin kidney protein 1, collectin sub-family member 11, collectin-11, DKFZp686N1868, MGC3279 | |
| Produced in rabbits immunized with purified, recombinant Mouse CL-K1/COLEC11 (Uniprot#: Q3SXB8-1; Met1-Leu272) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title