Learn More
Invitrogen™ CLAC-P Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA14307
Description
PA1-4307 detects CLAC-P (NC2-2 region) in human samples. PA1-4307 has been successfully used in Western blot procedures. By Western blot this antibody detects bands of 50, 70, and 100 kDa representing CLAC-P in formic acid extracts of Alzheimer's disease brain. PA1-4307 immunizing peptide corresponds to amino acid residues 155-169 of the NC2-2 region of human and mouse CLAC-P.
Alzheimer's disease (AD) is characterized by deposition of beta-amyloid as senile plaques and cerebral amyloid angiopathy. The principal component of AD amyloid is the amyloid beta peptide (A beta). Although production and deposition of A beta appear closely related to the pathogenesis of AD, current evidence suggest that A beta alone is not sufficient to cause neuronal death and symptoms of dementia. A component of senile plaque (SP), CLAC (collagen-like Alzheimer amyloid plaque component ) and its precursor CLAC-P are deposited with extracellular beta-amyloid and have been recently implicated in cell toxicity of beta-amyloid in AD brains. CLAC-P is a unique, membrane-bound collagen-like structure containing three Gly-X-Y repeat motifs and may define a novel class of neuronal collagens.
Specifications
| CLAC-P | |
| Polyclonal | |
| Unconjugated | |
| COL25A1 | |
| 2700062B08Rik; Alzheimer disease amyloid-associated protein; AMY; CFEOM5; CLAC; CLACP; CLAC-P; Col25a1; collagen alpha-1(XXV) chain; collagen type XXV alpha 1; collagen, type XXV, alpha 1; collagen-like Alzheimer amyloid plaque component; collagenous Alzheimer amyloid plaque component; procollagen, type XXV, alpha 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84570 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q9BXS0 | |
| COL25A1 | |
| Synthetic peptide: K(155) G E Q G D Q G P R M V F P K(169). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.