Learn More
MedChemExpress Cladribine
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Molecular Weight (g/mol) | 285.69 g/mol |
| Product Type | Reference compound |
| Content And Storage | 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light) |
| Shipping Condition | Room temperature |
| Purity | 98.92% |
| Quantity | 10 mg |
| Solubility Information | DMSO : ≥ 30 mg/mL (105.01 mM) ∣H2O : 10 mg/mL (35.00 mM∣Need ultrasonic) |
| For Use With (Application) | COVID-19-immunoregulation |
| CAS | 4291-63-8 |
| Synonym | 2-Chloro-2′-deoxyadenosine∣ CldAdo∣ 2CdA |