Learn More
OHAUS™ Clamp Connector
Product Code. 15702309 Shop All Ohaus Products
Change view
Click to view available options
Connector Types:
Bar
Channel
End-To-End
Hook
Lattice
Multi-Rod
Rod End
S
Material:
Aluminum
Nickel-plated
Stainless Steel
Size:
0 to 13 mm (Grip Range)
102 mm (L)
152 mm (L)
203 mm (L)
40 x 33 mm (H x L)
51 mm (L)
Unit Size:
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Material Size Unit Size
15702309 Rod End Nickel-plated 0 to 13 mm (Grip Range) Each
15702319 Channel Aluminum 51 mm (L) Each
15712309 S Nickel-plated 0 to 13 mm (Grip Range) Each
15712319 Channel Aluminum 102 mm (L) Each
15722309 Hook Nickel-plated 0 to 13 mm (Grip Range) Each
15722319 Bar Aluminum 152 mm (L) Each
15722409 Hook Stainless Steel 0 to 13 mm (Grip Range) Each
15732319 Bar Aluminum 203 mm (L) Each
15752309 Channel Nickel-plated 40 x 33 mm (H x L) Each
15762269 Rod End Stainless Steel 0 to 13 mm (Grip Range) Each
15762309 Multi-Rod Nickel-plated 40 x 33 mm (H x L) Each
15782309 End-To-End Aluminum 0 to 13 mm (Grip Range) Each
15792299 Lattice Aluminum 0 to 13 mm (Grip Range) Each
15792399 S Stainless Steel 0 to 13 mm (Grip Range) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 15702309 Supplier OHAUS™ Supplier No. 30392256
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
OHAUS™ Clamp Connector provides secure support for holding glassware at elevated positions.
  • Hold glassware safely
  • Corrosion-resistant
  • Providing convenience with exceptional stability and durability
TRUSTED_SUSTAINABILITY
Specifications
Connector Types Rod End
Type Connector
Material Nickel-plated
Includes Fixed 90° Connection Angle
Description Clamp, Connector, Rod End, CLC-RDENDZ
Size 0 to 13 mm (Grip Range)
Size (Metric) Grip 0 to 13 mm