Learn More
OHAUS™ Clamp Connector
Shop All Ohaus Products
Click to view available options
Connector Types:
Bar
Channel
End-To-End
Hook
Lattice
Multi-Rod
Rod End
S
Material:
Aluminum
Nickel-plated
Stainless Steel
Size:
0 to 13 mm (Grip Range)
102 mm (L)
152 mm (L)
203 mm (L)
40 x 33 mm (H x L)
51 mm (L)
Unit Size:
Each
Description
- Hold glassware safely
- Corrosion-resistant
- Providing convenience with exceptional stability and durability
Specifications
Specifications
| Connector Types | Channel |
| Type | Connector |
| Material | Aluminum |
| Description | Clamp, Connector, Bar, CLC-HTZMBA10 |
| Length (English) | 4 in. |
| Length (Metric) | 102 mm |
| Size | 102 mm (L) |