Learn More
Fisherbrand™ Cellulose Stoppers
For flasks, erlenmeyers and test tubes.
Brand:  Fisherbrand™
Additional Details : Weight : 0.83000kg
 View more versions of this product
Product Code. 11778513
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Permeable in air
  • Sterilizable to 200°C maximum
  • Single use
Specifications
Specifications
Yes
Stopper
8 mm
1000PK
200°C
Sterile
Beakers, Erlenmeyer flasks, tubes, and different types of bottles
16 mm
Cellulose Fiber
35 mm