Learn More
Fisherbrand™ Stainless Steel Forceps
With rubber-sheathed jaws for a soft and secure hold.
Brand:  Fisherbrand™
Additional Details : Weight : 0.14400kg
Product Code. 11855111
  • € 55.50 / Each
Estimated Shipment: 21-06-2024
to see stock.
Add to basket
Description
Description
  • 3-finger design
  • 80mm diameter opening
  • Rod diameter of 10mm, length of 135mm
Specifications
Specifications
1 Ea.
Clamp Rexaloy
Stainless Steel Pummels/Twin Screws