Learn More
OHAUS™ Clamp Ribbon
Product Code. 17161001 Shop All Ohaus Products
Click to view available options
Dimensions (L x W x H):
30.000 x 20.000 x 5.000 cm
Unit Size:
1 unit
Product Code. 17161001
Brand: OHAUS™ 30586774
Only null left
Add to basket
Add to basket
Clamp Ribbon for use with overhead stirrers.
  • Overhead Stirrer Accessory
TRUSTED_SUSTAINABILITY
Specifications
Height (Metric) 5.000 cm
Width (Metric) 20.000 cm
Weight (Metric) 330.000 g
Dimensions (L x W x H) 30.000 x 20.000 x 5.000 cm