Learn More
Kartell™ Clamps for Glass Joints
Click to view available options
Size:
10/19
34/35
45/40
Unit Size:
Pack of 10
Description
- Corrosion proof
- Do not scratch glass
- Available in various colours according to dimension
- Autoclavable
Specifications
Specifications
| Quantity | 10/Pk. |
| Material | POM |
| Size | 34/35 |
| Color | Orange |