Learn More
Kartell™ Clamps for Glass Joints
Product Code. p-7156834
Click to view available options
Size:
10/19
34/35
45/40
Unit Size:
Pack of 10
Product Code. 15875577
Brand: Kartell™ 3101900
Only null left
Add to basket
Add to basket
Plastic clamps for safe connection of standard-finish glass conical joints
  • Corrosion proof
  • Do not scratch glass
  • Available in various colours according to dimension
  • Autoclavable
Specifications
Quantity 10/Pk.
Material POM
Size 10/19
Color Black