Learn More
Clascoterone, MedChemExpress
Product Code. 16496166
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 16496166
Brand: Medchem Express HY13331
MedChemExpress Clascoterone (Cortexolone 17 alpha-propionate;Cortexolone 17α-propionate;CB-03-01) is a new topical and peripherally selective androgen antagonist.
Clascoterone (Cortexolone 17 alpha-propionate
Specifications
Chemical Name or Material Clascoterone
Physical Form Solid
CAS 19608-29-8
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C24H34O5
Synonym Cortexolone 17 alpha-propionate Cortexolone 17α-propionate CB-03-01
Solubility Information DMSO : ≥ 100 mg/mL (248.43 mM)
SMILES O=C1CC[C@@]2(C)C(CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@@]4(C(CO)=O)OC(CC)=O)=C1
Molecular Weight (g/mol) 402.52
Formula Weight 402.52
Percent Purity 98.21%
Grade Research
Show More Show Less
Research purposes only