missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLASP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38380-100ul
This item is not returnable.
View return policy
Description
CLASP1 Polyclonal antibody specifically detects CLASP1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| CLASP1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| CLIP-associating protein 1, cytoplasmic linker associated protein 1, Cytoplasmic linker-associated protein 1, DKFZp686D1968, DKFZp686H2039, hOrbit1, KIAA0622FLJ33821, MAST1FLJ41222, MGC131895, multiple asters 1, Multiple asters homolog 1, Protein Orbit homolog 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1389-1538 of human CLASP1 (NP_056097.1).,, Sequence:, IMKTLEAHKDSHKEVVRAAEEAASTLASSIHPEQCIKVLCPIIQTADYPINLAAIKMQTKVVERIAKESLLQLLVDIIPGLLQGYDNTESSVRKASVFCLVAIYSVIGEDLKPHLAQLTGSKMKLLNLYIKRAQTTNSNSSSSSDVSTHS | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23332 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction