missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CLASP1 Recombinant Rabbit Monoclonal Antibody (23GB3810), Invitrogen™

Product Code. 30088992
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30088992 20 μL 20µL
30087667 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30088992 Supplier Invitrogen™ Supplier No. MA553645

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

CLASPs, such as CLASP1, are nonmotor microtubule-associated proteins that interact with CLIPs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLASP1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3810
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CLASP1
Gene Accession No. Q7Z460, Q80TV8
Gene Alias 1700030C23Rik; 5730583A19Rik; B130045P17Rik; CLASP1; CLASP1alpha; CLIP associating protein 1; CLIP-associating protein 1; cytoplasmic linker associated protein 1; cytoplasmic linker-associated protein 1; hOrbit1; Kiaa0622; MAST1; multiple asters 1; multiple asters homolog 1; Protein Orbit homolog 1
Gene Symbols CLASP1
Host Species Rabbit
Immunogen A synthetic peptide of human CLASP1 (1-52AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23332, 304740, 76707
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.