Learn More
Invitrogen™ Claspin Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5102840
Product Code. 16659294
  • € 498.00 / 100µL
Description
Antibody detects endogenous levels of total Claspin.
Xenopus claspin is an essential upstream regulator of checkpoint kinase 1 and triggers a checkpoint arrest of the cell cycle in the presence of DNA templates in Xenopus egg extracts. The human gene appears to be the homolog Xenopus claspin and its function has not been determined.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Claspin
Polyclonal
Unconjugated
CLSPN
B130025E01; C85083; claspin; claspin homolog; CLSPN; E130314M08Rik; hClaspin; MGC131612; MGC131613; MGC131615
Rabbit
Affinity chromatography
RUO
269582, 298534, 63967
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q80YR7, Q9HAW4
CLSPN
A synthesized peptide derived from human CLSPN(Accession Q9HAW4), corresponding to amino acid residues H10-K60.
100 μL
Primary
Human, Mouse, Rat