missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLASRP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566494
This item is not returnable.
View return policy
Description
Immunogen sequence: PAVGEKLKKTE PAAGKETGAA KPKLTPQEKL KLRMQKALNR QFKADKKAAQ EKMIQQEHER QEREDELRAM Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.
CLASRP is probably functions as an alternative splicing regulator. CLASRP may regulate the mRNA splicing of genes such as CLK1. This may act by regulating members of the CLK kinase family.
Specifications
| CLASRP | |
| Polyclonal | |
| Unconjugated | |
| CLASRP | |
| Clasp; Clasrp; CLK4 associating serine/arginine rich protein; Clk4 associating SR-related protein; CLK4-associating serine/arginine rich protein; Clk4-associating SR-related protein; Serine/arginine-rich splicing factor 16; Sfrs16; Splicing factor, arginine/serine-rich 16; splicing factor, arginine/serine-rich 16 (suppressor-of-white-apricot homolog, Drosophila); Srsf16; suppressor of white apricot homolog 2; suppressor of white-apricot homolog 2; SWAP2; UNQ2428/PRO4988 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11129 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N2M8 | |
| CLASRP | |
| Recombinant Human CLASRP. Recombinant protein control fragment (Product #RP-107165). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction