Learn More
BRAND™ Class A Amber Borosilicate Volumetric Flask with Polypropylene Stopper
Product Code. 16471861 Shop All Brand Products
Click to view available options
:
Amber Volumetric Flask
Product Code. 16471861
Brand: BRAND™ 237483
Only null left
Add to basket
Add to basket
Amber volumetric flask with polypropylene stopper and USP batch certificate.
  • Borosilicate 3.3
  • To DIN EN ISO 1042
  • Calibrated to contain (TC, In)
  • Includes USP certificate single batch delivery
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (3.3)
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Narrow
Sterility Non-sterile
Bottom Shape Flat