Learn More
BRAND™ Class A Amber Borosilicate Volumetric Flask with Polypropylene Stopper
Shop All Brand Products
Click to view available options
:
Amber Volumetric Flask
Description
- Borosilicate 3.3
- To DIN EN ISO 1042
- Calibrated to contain (TC, In)
- Includes USP certificate single batch delivery
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Graduated | Yes |
| Mouth Style | Narrow |
| Sterility | Non-sterile |
| Bottom Shape | Flat |