missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Class A Amber Glass White Graduated Measuring Flasks

Product Code. 10776253
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
Unit Size:
Pack of 2
This item is not returnable. View return policy

Product Code. 10776253

Brand: Hirschmann™ 264018627

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Provided in amber glass for the protection of light-sensitive contents.

  • White graduation
  • Class A
  • With dated batch identification
  • Standard ground and PE stopper
  • Manufactured from DURAN™ borosilicate glass for high chemical and thermal resistance
  • USP certified

Specifications

Closure Material Polyethylene
Color Amber
Capacity (Metric) 250 mL
Material Borosilicate Glass
For Use With (Application) Liquid handling of light-sensitive products
Height (Metric) 220 ± 5 mm
Standard Taper Joint Ground 14/23
Graduated Yes
Class A
Bottom Shape Flat
Closure Type Stopper
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.