missing translation for 'onlineSavingsMsg'
Learn More

Witeg™ Class A Conformity Certified Volumetric Flasks

Product Code. 10134394
Change view
Click to view available options
Capacity (Metric):
25 mL
300 mL
5 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10134394 300 mL Each
10086281 25 mL Each
10161571 5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10134394 Supplier Witeg™ Supplier No. 3 664 030
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Provided with batch certificate in the box.

  • Inclusive batch certificate
  • Made from DURAN™ tubing
  • Brown graduation
  • With stoppers of PE
  • A range of capacities and tolerances available

Specifications

Closure Material Polyethylene
Capacity (Metric) 300 mL
Material Glass
Graduations Brown
Tolerance ±0.150 mL
Graduated Yes
Class A
Closure Type Stopper
Closure Size NS 14/23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.