Learn More
Witeg™ Class A Conformity Certified Volumetric Flasks
Product Code. 10161571
Change view
Click to view available options
Capacity (Metric):
25 mL
300 mL
5 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10161571 5 L Each
10086281 25 mL Each
10134394 300 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10161571 Supplier Witeg™ Supplier No. 3 664 500
Provided with batch certificate in the box.
  • Inclusive batch certificate
  • Made from DURAN™ tubing
  • Brown graduation
  • With stoppers of PE
  • A range of capacities and tolerances available
Specifications
Closure Material Polyethylene
Capacity (Metric) 5 L
Material Glass
Graduations Brown
Tolerance ±1.200 mL
Graduated Yes
Class A
Closure Type Stopper
Closure Size NS 34/35