Learn More
Hirschmann™ AR™ Glass Volumetric Pipettes, Class AS
Click to view available options
Volume (Metric):
0.5 mL
1 mL
2 mL
2.5 mL
3 mL
5 mL
10 mL
15 mL
20 mL
25 mL
50 mL
100 mL
Unit Size:
Pack of 12
Pack of 6
Description
- Acc. to DIN EN ISO 648
- One mark, blue graduation
- Conformity certified, with dated batch identification
Specifications
Specifications
| Accuracy | ± 0.005 mL |
| Autoclavable | Autoclavable |
| Calibration | To Deliver |
| Certifications/Compliance | DIN EN ISO 648 |
| Class | AS |
| Material | Soda Lime Glass |
| Sterility | Non-sterile |
| Graduated | Yes |
| Graduation Color | Blue |
| Graduation Type | Blue enamel color |
| Show More |