Learn More
Hirschmann™ Class AS Volumetric Pipettes with Amber Stain Graduation
Click to view available options
Volume (Metric):
0.5 mL
1 mL
1.5 mL
2 mL
2.5 mL
3 mL
4 mL
5 mL
6 mL
7 mL
8 mL
9 mL
10 mL
15 mL
20 mL
25 mL
30 mL
40 mL
50 mL
100 mL
Unit Size:
Pack of 12
Pack of 6
Description
- One mark
- Acc. to DIN EN ISO 648
- Conformity certified, with dated batch identification
Specifications
Specifications
| Class | AS |
| Material | AR-GLAS |
| Graduated | Yes |
| Graduation Color | Brown |
| Quantity | 12/Pk. |
| Tolerance | 0.02 mL |
| Volume (Metric) | 8 mL |
| Length (Metric) | 450 mm |
| Type | Volumetric Pipet |