Learn More
Hirschmann™ Class AS Graduated Pipettes with Amber Graduation
Click to view available options
Volume (Metric):
0.5 mL
1 mL
2 mL
5 mL
10 mL
20 mL
Graduations:
0.01 mL
0.02 mL
0.05 mL
0.1 mL
Unit Size:
Pack of 12
Pack of 6
Description
- Main point ring graduation
- Graduated not to tip, zero on top
- Acc. to DIN EN ISO 835
- Conformity certified, with dated batch identification
Specifications
Specifications
| Accuracy | 0.01 mL |
| Autoclavable | Autoclavable |
| Calibration | To Deliver |
| Certifications/Compliance | DIN EN ISO 835 |
| Class | AS |
| Material | Soda Lime Glass |
| Sterility | Non-sterile |
| Graduated | Yes |
| Graduation Color | Amber |
| Graduations | 0.1 mL |
| Show More |