Learn More
Witeg™ Class AS Graduated Volumetric Pipettes
Tempered delivery tips offering a wear resistance which is up to 7 times better than that of conventional pipette delivery tips.
€ 4.45 - € 9.55
Specifications
Certifications/Compliance | Conformity certified; DIN EN ISO 385 |
---|---|
Class | AS |
Material | Glass |
Graduated | Yes |
Graduation Color | Amber |
Product Code | Brand | Volume (Metric) | Color | Graduations | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Color | Graduations | Price | Quantity & Availability | |||
10214061
|
Witeg™
3 510 810 |
20 mL | Yellow | 0.1 mL divisions |
€ 9.55
Each |
In Stock
Log in to see stock. |
|||
10674392
|
Witeg™
3 510 011 |
0.1 mL | Green | 0.001 mL divisions |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Conformity certified
- Main point ring graduation
- With ISO color code
- Acc. to DIN ENO ISO 385
- With amber stain graduations and inscriptions
- Sizes 5 mL and up with construction in the suction to hold cotton plug
- Waiting time of 5 seconds
- For complete and swift delivery
Specifications
Conformity certified; DIN EN ISO 385 | |
Glass | |
Amber | |
Tempered Delivery |
AS | |
Yes | |
Main point ring | |
Graduated Pipet |