Learn More
Hirschmann™ Class B PMP Volumetric Flask with Stopper
Click to view available options
Capacity (Metric):
10 mL
50 mL
500 mL
1000 mL
Unit Size:
Pack of 3
Pack of 4
Pack of 6
Description
- Acc. to DIN EN ISO 1042
- PMP (TPX)
- Adjusted ringmark
- Translucent
- With standard ground poly stoppers
Specifications
Specifications
| Certifications/Compliance | DIN EN ISO 1042 |
| Closure Material | PMP |
| Capacity (Metric) | 10 mL |
| Height (Metric) | 90 mm |
| No. per Pack | 6/Pack |
| Tolerance | ± 0.08 mL |
| Class | B |
| Bottom Shape | Round |
| Closure Type | Stopper |
| Closure Size | NS 10/19 |