Learn More
VITLAB™ Class B Polypropylene Graduated Cylinders with Raised Blue Scale
Use these cylinders for food industry and volume measurement applications. VITLAB™ Class B Polypropylene Graduated Cylinders with Raised Blue Scale feature easy to read, raised, embossed blue scale and ring marks at the primary scale points.
Brand: VITLAB™ 648081
Additional Details : Weight : 0.01000kg
Description
- Highly transparent polypropylene graduated cylinder
- Hexagonal base with bottom studs provides high stability
- Calibrated (In)
- Class B tolerances according to DIN 12681 / ISO 6706.
- Thermal stress up to 80°C does not cause tolerance limits to be permanently exceeded
- To preserve markings, do not clean at temperatures exceeding 60°C
Specifications
Polypropylene | |
DIN 12681, ISO 6706 | |
Class B | |
1 mL | |
Round | |
Blue | |
200 mm |
±1 mL | |
6Pack | |
27 mm | |
±1mL | |
Food Industry and Volume Measurement | |
Hexagonal | |
Graduated Cylinder |