Learn More
VITLAB™ Class B Polypropylene Short Form Measuring Cylinders
Product Code. 11853662
Click to view available options
Graduations:
0.5 mL
1 mL
2 mL
5 mL
10 mL
20 mL
Unit Size:
Each
Pack of 12
Pack of 6
Product Code. 11853662
Brand: VITLAB™ 640941
Only null left
Add to basket
Add to basket
Use these cylinders for food industry and volume measurement applications. VITLAB™ Class B Polypropylene Short Form Measuring Cylinders feature easy to read, raised scale and ring marks at the primary scale points.
  • Highly transparent polypropylene graduated cylinder
  • Calibrated (In)
  • Class B tolerances according to DIN 12681 / ISO 6706.
  • Thermal stress up to 80°C does not cause tolerance limits to be permanently exceeded
Specifications
Class B
Graduations 0.5 mL
Limit of Error ±0.5mL
Shape Round
Height (Metric) 122 mm
Type Graduated Cylinder