Learn More
VITLAB™ Class B Polypropylene Short Form Measuring Cylinders
Click to view available options
Graduations:
0.5 mL
1 mL
2 mL
5 mL
10 mL
20 mL
Unit Size:
Each
Pack of 12
Pack of 6
Description
- Highly transparent polypropylene graduated cylinder
- Calibrated (In)
- Class B tolerances according to DIN 12681 / ISO 6706.
- Thermal stress up to 80°C does not cause tolerance limits to be permanently exceeded
Specifications
Specifications
| Material | Polypropylene |
| Tolerance | ±1 mL |
| Class | B |
| Diameter (Metric) | 27 mm |
| Graduations | 1 mL |
| Limit of Error | ±1mL |
| Shape | Round |
| Height (Metric) | 142 mm |
| Type | Graduated Cylinder |