Learn More
Zwiebel™ Plastic Box with Stainless Steel 25kg Weight, F1 with Certificate
Product Code. 11850922
Click to view available options
:
Mass: 20kg
Product Code. 11850922
Brand: Zwiebel™ PBI5682N
Only null left
Add to basket
Add to basket
Use with weighing instruments of special accuracy class I and high accuracy class II. Zwiebel™ Class F1 Weights are intended for use in the verification or calibration of class F2 weights.
  • Class F1 calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
Specifications
Product Type Box Weight
Classification F1
Mass 25 kg
For Use With (Application) Weighing Instruments of Special Accuracy Class I and High Accuracy Class II