Learn More
Zwiebel™ Class F1 Box Weights
Product Code. 12051633
Click to view available options
Mass:
50 mg
200 mg
500 mg
2 g
50 g
100 g
200 g
1000 g
2000 g
Tolerance:
±0.04 mg
±0.06 mg
±0.08 mg
±0.12 mg
±0.30 mg
±0.50 mg
±1.00 mg
±10 mg
±5.00 mg
Unit Size:
Each
Product Code. 12051633
Brand: Zwiebel™ PBI23075
Only null left
Add to basket
Add to basket
Use with weighing instruments of medium accuracy class II. Zwiebel™ Class F1 Box Weights are ideal for the verification or calibration of class F2 weights.
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Mass 2 g
Tolerance ±0.12 mg
For Use With (Application) For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II