Learn More
Zwiebel™ Class F1 Box Weights with Certificate
Click to view available options
Tolerance:
±0.1 mg
±0.20 mg
±0.25 mg
±0.30 mg
±0.50 mg
±25 mg
±5.0 mg
±50 mg
Unit Size:
Each
Description
- Class F1 calibration weight
- Delivered with a calibration certificate guaranteeing traceability
- In compliance with OIML R111
Specifications
Specifications
| Mass | 10,000 g |
| Tolerance | ±50 mg |
| For Use With (Application) | For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II |