Learn More
Zwiebel™ Class F1 Weight with Certificate
Product Code. 11750613
Click to view available options
:
Weight: 1 mg
Product Code. 11750613
Brand: Zwiebel™ PBI2301L
Only null left
Add to basket
Add to basket
Zwiebel™ F1 Weight with Certificate is intended for use in the verification or calibration of class F2 weights.
  • Permanent magnetization: < 25 μT
  • Weights intended for use in the verification or calibration of class F2 weights and for use with weighing instruments of special accuracy class I and high accuracy class II
  • One bloc material
Specifications
Product Type Wire Weight
Weight (Metric) 1 mg
Density 8500 ±100 kg/m3
Shape Triangle
Diameter (Metric) Outside 0.1 mm wire
Length (Metric) 4.5 mm short ends, 6.5 mm long end
Includes Certificate