Learn More
Zwiebel™ Class M1 Box Weights with Certificate
Click to view available options
Tolerance:
±1 mg
±1.2 mg
±1.6 mg
±10 mg
±100 mg
±2 mg
±250 mg
±3 mg
±5 mg
±500 mg
Unit Size:
Each
Description
- Class M1 stainless steel calibration weight
- Delivered with a calibration certificate guaranteeing traceability
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
| Tolerance | ±5 mg |
| For Use With (Application) | For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII |