Learn More
Zwiebel™ Class M1 Weight Boxes with Certificate
Click to view available options
Mass:
500 g
2000 g
Unit Size:
Each
Specifications
Specifications
| Mass | 500 g |
| Tolerance | ±25 mg |
| For Use With (Application) | For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII |