missing translation for 'onlineSavingsMsg'
Learn More

Zwiebel™ Class M2 Weight Set

Product Code. 11800962
Click to view available options
Mass:
1 to 100 g
1 to 200 g
1 to 50 g
1 to 500 g
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11800962

Brand: Zwiebel™ PBI56842

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M2 Weights are ideal for the verification or calibration of class M3 weights.

  • Class M2 stainless steel calibration weight
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box

Specifications

Mass 1 to 500 g
For Use With (Application) For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.