Learn More
Zwiebel™ Class M2 Weight Set
Product Code. 11840932
Click to view available options
Mass:
1 to 100 g
1 to 200 g
1 to 50 g
1 to 500 g
Unit Size:
Each
Product Code. 11840932
Brand: Zwiebel™ PBI56833
Only null left
Add to basket
Add to basket
Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M2 Weights are ideal for the verification or calibration of class M3 weights.
  • Class M2 stainless steel calibration weight
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Mass 1 to 50 g
For Use With (Application) For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II