missing translation for 'onlineSavingsMsg'
Learn More

Huenersdorf™ CLASSIC™ 8-Compartment Polypropylene Assortment Box

Product Code. 10729645
Change view
Click to view available options
Compartments:
8
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Compartments unitSize
10729645 8 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10729645 Supplier Huenersdorf™ Supplier No. 611700
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

These assortment boxes are manufactured from fracture-proof and dimensionally stable polypropylene with eight compartments.

  • Transparent polypropylene lid, red polypropylene base
  • With stable hinges and closures
  • Resistant to greases, resins, alkalis, oils and acids
  • Ideal for packaging purposes
  • Food-safe and temperature resistant from −20° to +70°C
  • With 8 compartments of varying sizes

Specifications

Closure Type Hinged lid
Color Transparent Lid, Red Base
Compartments 8
Dimensions (L x W x H) 225 x 335 x 55 mm
For Use With (Application) Packaging
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.