Learn More
Huenersdorf™ CLASSIC-FLEX™ Assortment Boxes
Product Code. 10609905
Change view
Click to view available options
Dimensions (L x W x H):
225 x 335 x 32 mm
225 x 335 x 73 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
10609905 225 x 335 x 32 mm Each
10011774 225 x 335 x 73 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10609905 Supplier Huenersdorf™ Supplier No. 613200
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
These assortment boxes are manufactured of extra dimensionally stable transparent polystyrene for individual fitting with insert boxes.
  • Transparent lid and bottom for optimum display of contents
  • With stable hinges and closures
  • Ideal for packaging purposes
  • Food-safe and temperature resistant from −20° to +70°C
  • Without compartment subdivisions for individual fitting with insert boxes
Specifications
Closure Color Transparent
Color Transparent
Material Polystyrene
Shape Rectangular
For Use With (Application) Food Safe
For Use With (Equipment) Insert boxes
Includes Stable Hinge and Closure
Product Type Assortment Box
Temperature Range (Metric) -20° to +70°C
Dimensions (L x W x H) 225 x 335 x 32 mm
Length (Metric) 225 mm
Width (Metric) 335 mm
Height (Metric) 32 mm