Learn More
Tenak™ Classic Rack for Chest Freezers, 75 mm Box Height, with Boxes
Shop All TENAK Products
Click to view available options
Holds:
Boxes
Unit Size:
Each
Description
- Comes with a locking rod to secure the cryoboxes
- Measure the inside height of you freezer when choosing the right rack
- Constructed of stainless steel
- Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications
Specifications
| Holds | Boxes |
| Material | Stainless Steel |
| Positions | 2 |
| For Use With (Equipment) | Chest Frseezers |
| Dimensions (L x W x H) | 140 x 140 x 166 mm |
| Includes | White cardboard boxes, 9 x 9 cell dividers |
| Height (Metric) | 75 mm (Box) |