missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clathrin Heavy Chain 1/CHC17 Antibody (CHC/1432), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08333AF405
This item is not returnable.
View return policy
Description
Clathrin Heavy Chain 1/CHC17 Monoclonal antibody specifically detects Clathrin Heavy Chain 1/CHC17 in Human, Mouse, Rat, Porcine, Avian, Bovine, Canine, Monkey, Xenopus samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| Clathrin Heavy Chain 1/CHC17 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| CHC, CHC17, clathrin heavy chain 1, Clathrin heavy chain on chromosome 17, clathrin, heavy chain, clathrin, heavy chain (Hc), clathrin, heavy polypeptide (Hc), clathrin, heavy polypeptide-like 2, CLH17, CLH-17, CLTCL2, Hc, KIAA0034 | |
| Recombinant full-length human Clathrin Heavy Chain 1/CHC17 protein | |
| 100 μg | |
| Membrane Trafficking and Chaperones, Membrane Vesicle Markers | |
| 1213 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunofluorescence | |
| CHC/1432 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Avian | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction