Learn More
Clathrin Heavy Chain 1/CHC17 Antibody (CLTC/1431), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-53255-100UG
 View more versions of this product
Product Code. 18297160
  • € 518.00 / 100µg
Description
Clathrin Heavy Chain 1/CHC17 Monoclonal specifically detects Clathrin Heavy Chain 1/CHC17 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clathrin Heavy Chain 1/CHC17
Monoclonal
0.2 mg/mL
Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, SDS-Page
CHC, CHC17, clathrin heavy chain 1, Clathrin heavy chain on chromosome 17, clathrin, heavy chain, clathrin, heavy chain (Hc), clathrin, heavy polypeptide (Hc), clathrin, heavy polypeptide-like 2, CLH17, CLH-17, CLTCL2, Hc, KIAA0034
Mouse
192 kDa
100 μg
Membrane Trafficking and Chaperones, Membrane Vesicle Markers
1213
Store at 4C.
IgG1 κ
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page
CLTC/1431
Unconjugated
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
CLTC
Purified recombinant N-terminal fragment of human Clathrin Heavy Chain (exact sequence is proprietary)
Protein A or G purified