missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clathrin Heavy Chain Antibody (Academia Sinica patent anti-Clathrin Heavy Chain), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28577CL1
This item is not returnable.
View return policy
Description
Clathrin Heavy Chain Monoclonal antibody specifically detects Clathrin Heavy Chain in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Clathrin Heavy Chain | |
| Monoclonal | |
| CoraFluor 1 | |
| Human | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| Academia Sinica patent anti-Clathrin Heavy Chain | |
| PBS | |
| Clathrin Heavy Chain/ CHC | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction