missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clathrin Heavy Chain Antibody (Academia Sinica patent anti-Clathrin Heavy Chain) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28577JF525
This item is not returnable.
View return policy
Description
Clathrin Heavy Chain Monoclonal antibody specifically detects Clathrin Heavy Chain in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Clathrin Heavy Chain | |
| Monoclonal | |
| Janelia Fluor 525 | |
| Human | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| Academia Sinica patent anti-Clathrin Heavy Chain | |
| 50mM Sodium Borate | |
| Clathrin Heavy Chain/ CHC | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction