Learn More
Invitrogen™ Clathrin Heavy Chain Monoclonal Antibody (X22), Alexa Fluor™ 555

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA1065A555
Description
MA1065A555 detects clathrin heavy chain in non-human primate, bovine, human, rat and mouse tissues as well as hamster (CHO) cells. The MA1065A555 immunogen is purified human brain clathrin heavy chain. Electron microscopy and proteolysis mapping demonstrate that MA1065A555 binding occurs towards the central hub of the triskelion, N-terminal to the light chain binding regions.
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
Specifications
| Clathrin Heavy Chain | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
| P11442, P49951, P53675, Q00610, Q68FD5 | |
| CLTC, CLTCL1 | |
| Purified human brain clathrin heavy chain. | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey, Bovine, Hamster | |
| Antibody | |
| IgG1 |
| Immunocytochemistry | |
| X22 | |
| Alexa Fluor 555 | |
| CLTCL1 | |
| 3110065L21Rik; CHC; CHC17; CHC22; clathrin heavy chain; clathrin heavy chain 1; Clathrin heavy chain 2; clathrin heavy chain like 1; clathrin heavy chain on chromosome 17; Clathrin heavy chain on chromosome 22; clathrin, heavy chain (Hc); clathrin, heavy chain-like 1; clathrin, heavy polypeptide (Hc); Clathrin, heavy polypeptide D; clathrin, heavy polypeptide-like 1; clathrin, heavy polypeptide-like 2; CLH17; CLH-17; CLH22; CLH-22; Cltc; CLTCL; CLTCL1; CLTCL2; CLTD; Hc; KIAA0034; R74732 | |
| Mouse | |
| Protein A | |
| RUO | |
| 100756644, 1213, 281080, 54241, 67300, 8218 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.