missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clathrin interactor 1 Antibody (OTI8C1) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71491
This item is not returnable.
View return policy
Description
Clathrin interactor 1 Monoclonal specifically detects Clathrin interactor 1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Clathrin interactor 1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000 | |
| clathrin interacting protein localized in the trans-Golgi region, clathrin interactor 1, Clathrin-interacting protein localized in the trans-Golgi region, CLINT, ENTHClint, enthoprotin, EPN4Enthoprotin, EPNRepsin 4, Epsin-4, EpsinR, Epsin-related protein, FLJ46753, KIAA0171epsinR | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| OTI8C1 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| CLINT1 | |
| Human recombinant protein fragment corresponding to amino acids 1-290 of human CLINT1(NP_055481) produced in E.coli. | |
| 100 μg | |
| 9685 | |
| Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction