missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clathrin light chain Antibody (CLC/1421), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54436H
This item is not returnable.
View return policy
Description
Clathrin light chain Monoclonal antibody specifically detects Clathrin light chain in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Clathrin light chain | |
| Monoclonal | |
| HRP | |
| clathrin light chain A, clathrin, light chain A, LCA, light polypeptide (Lca) | |
| Purified recombinant N-terminal fragment of human Clathrin Light Chain | |
| 100 μg | |
| Membrane Trafficking and Chaperones, Membrane Vesicle Markers | |
| 1211 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CLC/1421 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction