Learn More
Claudin-1 Antibody (421203), Allophycocyanin/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB4618APCCY7
This item is not returnable.
View return policy
Description
Claudin-1 Monoclonal antibody specifically detects Claudin-1 in Human samples. It is validated for Flow Cytometry
Specifications
| Claudin-1 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 421203 | |
| Flow Cytometry | |
| claudin 1, claudin-1, CLD1, ILVASCSenescence-associated epithelial membrane protein, SEMP1senescence-associated epithelial membrane protein 1 | |
| CHO Chinese hamster ovary cell line transfected with human Claudin-1, Met1-Val211, Accession # O95832 | |
| 0.1 mL | |
| Extracellular Matrix | |
| 9076 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |